Skip to Content
MilliporeSigma
All Photos(1)

Documents

V900016

Sigma-Aldrich

L-(+)-Lactic acid

Vetec, reagent grade, 86%

Synonym(s):

(S)-2-Hydroxypropionic acid, Sarcolactic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6O3
CAS Number:
Molecular Weight:
90.08
Beilstein:
1720251
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

86%

SMILES string

C[C@H](O)C(O)=O

InChI

1S/C3H6O3/c1-2(4)3(5)6/h2,4H,1H3,(H,5,6)/t2-/m0/s1

InChI key

JVTAAEKCZFNVCJ-REOHCLBHSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1C

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Franco Valenza et al.
Critical care (London, England), 9(6), 588-593 (2005-12-17)
Lactate measurement in the critically ill has been traditionally used to stratify patients with poor outcome. However, plasma lactate levels are the result of a finely tuned interplay of factors that affect the balance between its production and its clearance.
J K Nicholson et al.
The Biochemical journal, 211(3), 605-615 (1983-06-01)
Spin echo Fourier transform proton n.m.r. spectra of whole blood contain resonances from both erythrocytes and plasma. A large number of well-resolved signals from mobile protons of low-molecular-weight metabolites in plasma and serum have been identified. Spectra from the plasmas
T Isotalo et al.
BJU international, 88(1), 30-34 (2001-07-12)
To assess whether patients in acute urinary retention from benign prostatic enlargement can be treated with a combined therapy comprising finasteride and a bioabsorbable self-reinforced poly L-lactic acid (SR-PLLA) urethral stent. Fifty-five patients in acute urinary retention were treated as
S Walenta et al.
Current medicinal chemistry, 11(16), 2195-2204 (2004-07-29)
A number of studies have demonstrated that malignant transformation is associated with an increase in glycolytic flux and in anaerobic and aerobic cellular lactate excretion. Using quantitative bioluminescence imaging in various primary carcinomas in patients (uterine cervix, head and neck
Souhaila Bouatra et al.
PloS one, 8(9), e73076-e73076 (2013-09-12)
Urine has long been a "favored" biofluid among metabolomics researchers. It is sterile, easy-to-obtain in large volumes, largely free from interfering proteins or lipids and chemically complex. However, this chemical complexity has also made urine a particularly difficult substrate to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service