Skip to Content
MilliporeSigma
All Photos(1)

Documents

V800365

Sigma-Aldrich

Sodium acetate trihydrate

AR, 99-102%

Synonym(s):

Acetic acid sodium salt trihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COONa · 3H2O
CAS Number:
Molecular Weight:
136.08
Beilstein:
3732037
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

AR

product line

Vetec

Assay

99-102%

form

crystalline powder

autoignition temp.

1112 °F

pH

8.5-9.5 (5% in water)

anion traces

chloride (Cl-): ≤0.0005%

cation traces

Fe: ≤0.0005%
heavy metals (as Pb): ≤0.0005%

SMILES string

O.O.O.[Na+].CC([O-])=O

InChI

1S/C2H4O2.Na.3H2O/c1-2(3)4;;;;/h1H3,(H,3,4);;3*1H2/q;+1;;;/p-1

InChI key

AYRVGWHSXIMRAB-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Other Notes

Enzyme substrate for acetate kinase (EC 2.7.2.1)

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Claire E Mason et al.
Journal of the American Society for Mass Spectrometry, 24(2), 274-285 (2013-01-04)
Single-stranded DNA-binding proteins (SSBs) are ubiquitous oligomeric proteins that bind with very high affinity to single-stranded DNA and have a variety of essential roles in DNA metabolism. Nanoelectrospray ionization mass spectrometry (nanoESI-MS) was used to monitor subunit exchange in full-length
Horng-Ruey Chua et al.
Journal of critical care, 27(2), 138-145 (2012-03-24)
The purpose of the study was to determine the effects of Plasma-Lyte 148 (PL) vs 0.9% saline (NS) fluid resuscitation in diabetic ketoacidosis (DKA). A multicenter retrospective analysis of adults admitted for DKA to the intensive care unit, who received
Rishiram Ramanan et al.
FEBS letters, 587(4), 370-377 (2013-01-15)
Phenotypic and genotypic changes in Chlamydomonas reinhardtii BafJ5, a starchless mutant, with respect to lipid metabolism was studied in different trophic states under nitrogen (N) sufficient and limited conditions. Interestingly, cellular lipid content increased linearly with input acetate concentration with
Qiong Jin et al.
Bioresource technology, 117, 148-154 (2012-05-23)
Dietzia natronolimnaea JQ-AN was isolated from industrial wastewater containing aniline. Under aerobic conditions, the JQ-AN strain degraded 87% of the aniline in a 300 mg L(-1) aniline solution after 120 h of shake flask incubation in a medium containing sodium
S Srinivasa et al.
The British journal of surgery, 100(1), 66-74 (2012-11-08)
Goal-directed fluid therapy (GDFT) has been compared with liberal fluid administration in non-optimized perioperative settings. It is not known whether GDFT is of value within an enhanced recovery protocol incorporating fluid restriction. This study evaluated GDFT under these circumstances in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service