Skip to Content
MilliporeSigma
All Photos(1)

Documents

V800360

Sigma-Aldrich

Silver sulfate

AR, ≥99%

Synonym(s):

Sulfuric acid disilver(I) salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ag2SO4
CAS Number:
Molecular Weight:
311.80
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

AR

product line

Vetec

Assay

≥99%

form

crystalline powder

pH

4.5-5 (0.5% in water)

mp

652 °C (lit.)

anion traces

chloride (Cl-): ≤0.005%

cation traces

Fe: ≤0.001%
heavy metals (as Pb): ≤0.0005%

SMILES string

[Ag+].[Ag+].[O-]S([O-])(=O)=O

InChI

1S/2Ag.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

YPNVIBVEFVRZPJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service