Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V800233

Sigma-Aldrich

Lead(II) acetate basic

LR, anhydrous, 72-75% Pb basis, 21.5-24% acetic acid basis

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
grade:
LR
anhydrous
form:
powder
Pricing and availability is not currently available.

grade

LR
anhydrous

product line

Vetec

form

powder

composition

acetic acid, 21.5-24%
Pb, 72-75%

SMILES string

O[PbH2]O.CC(=O)O[PbH2]OC(C)=O

InChI

1S/2C2H4O2.2H2O.2Pb/c2*1-2(3)4;;;;/h2*1H3,(H,3,4);2*1H2;;/q;;;;2*+2/p-4

InChI key

ODPICMBMWOLHAJ-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Lact. - Repr. 1A - STOT RE 1

Target Organs

Central nervous system,Blood,Immune system,Kidney,Reproductive organs

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service