Skip to Content
MilliporeSigma
All Photos(1)

Documents

V800129

Sigma-Aldrich

Copper(II) chloride

LR, ≥98%

Synonym(s):

Cupric chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CuCl2
CAS Number:
Molecular Weight:
134.45
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

LR

product line

Vetec

Assay

≥98%

form

powder

mp

620 °C (lit.)

density

3.386 g/mL at 25 °C (lit.)

SMILES string

Cl[Cu]Cl

InChI

1S/2ClH.Cu/h2*1H;/q;;+2/p-2

InChI key

ORTQZVOHEJQUHG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kenji Funaki et al.
Organic letters, 14(24), 6186-6189 (2012-12-12)
Direct arylation of thiophenes and benzothiophenes with aryltrimethylsilanes was effectively catalyzed by PdCl(2)(MeCN)(2) in the presence of CuCl(2) as an oxidant. The reaction preferentially occurred at the β-position of both thiophenes and benzothiophenes.
Mónica João Barros Amorim et al.
Environmental pollution (Barking, Essex : 1987), 164, 164-168 (2012-03-01)
Environmental effects of copper nanoparticles are little studied in terrestrial ecosystems. In the present article, the toxicity of copper nanoparticles (Cu-NP) on the enchytraeid Enchytraeus albidus is compared to the toxicity of a copper-salt (CuCl(2)). The effect parameters studied were
Iron(II)-catalyzed trifluoromethylation of potassium vinyltrifluoroborates.
Andrew T Parsons et al.
Angewandte Chemie (International ed. in English), 51(12), 2947-2950 (2012-02-14)
Francesca Cima et al.
Chemosphere, 89(1), 19-29 (2012-05-01)
After the widespread ban of TBT, due to its severe impact on coastal biocoenoses, mainly related to its immunosuppressive effects on both invertebrates and vertebrates, alternative biocides such as Cu(I) salts and the triazine Irgarol 1051, the latter previously used
Johnson J Liu et al.
Biochemical pharmacology, 85(2), 207-215 (2012-11-06)
Dorsal root ganglion (DRG) neurons are affected by platinum-induced neurotoxicity and neurodegenerative processes associated with disturbed copper homeostasis and transport. This study aimed to understand the role of copper transporter 1 (Ctr1) in the uptake and toxicity of copper and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service