Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V800101

Sigma-Aldrich

Calcium carbonate

AR, ≥98.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaCO3
CAS Number:
Molecular Weight:
100.09
Beilstein:
8008338
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
grade:
AR
form:
powder
Pricing and availability is not currently available.

grade

AR

product line

Vetec

Assay

≥98.5%

form

powder

pH

8

density

2.93 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.005%
sulfate (SO42-): ≤0.005%

cation traces

Ba: ≤0.01%
Cu: ≤0.0005%
Fe: ≤0.001%
K: ≤0.005%
Mg: ≤0.02%
Na: ≤0.10%
heavy metals (as Pb): ≤0.0005%

SMILES string

[Ca++].[O-]C([O-])=O

InChI

1S/CH2O3.Ca/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2

InChI key

VTYYLEPIZMXCLO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jane A Cauley et al.
Journal of women's health (2002), 22(11), 915-929 (2013-10-18)
Clinical outcomes of the Women's Health Initiative (WHI) calcium plus vitamin D supplementation trial have been reported during 7.0 years of active intervention. We now report outcomes 4.9 years after the intervention stopped and cumulative findings. Postmenopausal women (N=36,282) were
M Azizur Rahman et al.
PloS one, 8(3), e58781-e58781 (2013-03-26)
An organic matrix consisting of a protein-polysaccharide complex is generally accepted as an important medium for the calcification process. While the role this "calcified organic matrix" plays in the calcification process has long been appreciated, the complex mixture of proteins
Julyan H E Cartwright et al.
Angewandte Chemie (International ed. in English), 51(48), 11960-11970 (2012-11-06)
Although the polymorphism of calcium carbonate is well known, and its polymorphs--calcite, aragonite, and vaterite--have been highly studied in the context of biomineralization, polyamorphism is a much more recently discovered phenomenon, and the existence of more than one amorphous phase
Christopher T Parsons et al.
Environmental pollution (Barking, Essex : 1987), 178, 254-263 (2013-04-17)
Arsenic contamination of floodplain soils is extensive and additional fresh arsenic inputs to the pedosphere from human activities are ongoing. We investigate the cumulative effects of repetitive soil redox cycles, which occur naturally during flooding and draining, on a calcareous
Pedram Fatehi et al.
Carbohydrate polymers, 94(1), 531-538 (2013-04-03)
In this work, we aimed at adsorbing the oligo-sugars of prehydrolysis liquor on precipitated calcium carbonate (PCC) to produce modified PCC. The results showed that the adsorptions of oligo-sugars, lignin and furfural were greater on porous PCC (PCC2) than on

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service