Skip to Content
MilliporeSigma
All Photos(1)

Documents

V800036

Sigma-Aldrich

Ammonium hydrogen difluoride

AR, ≥95%

Synonym(s):

Ammonium bifluoride, Etching powder

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4HF2
CAS Number:
Molecular Weight:
57.04
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

AR

product line

Vetec

Assay

≥95%

form

crystals

mp

125 °C (lit.)

anion traces

chloride (Cl-): ≤0.002%

cation traces

Fe: ≤0.005%
heavy metals (as Pb): ≤0.01%

SMILES string

N.F.F

InChI

1S/2FH.H3N/h2*1H;1H3

InChI key

KVBCYCWRDBDGBG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Skin Corr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sebastian Ribi et al.
Oncotarget, 6(10), 7727-7740 (2015-03-13)
Somatic mutations of TP53 are among the most common in cancer and germline mutations of TP53 (usually missense) can cause Li-Fraumeni syndrome (LFS). Recently, recurrent genomic rearrangements in intron 1 of TP53 have been described in osteosarcoma (OS), a highly
Jinhong Li et al.
The New phytologist, 208(1), 137-148 (2015-04-14)
Heteromorphic flower development in Primula is controlled by the S locus. The S locus genes, which control anther position, pistil length and pollen size in pin and thrum flowers, have not yet been characterized. We have integrated S-linked genes, marker
Hiroko Sano et al.
PLoS genetics, 11(5), e1005209-e1005209 (2015-05-29)
The coordination of growth with nutritional status is essential for proper development and physiology. Nutritional information is mostly perceived by peripheral organs before being relayed to the brain, which modulates physiological responses. Hormonal signaling ensures this organ-to-organ communication, and the
Nick Orr et al.
Human molecular genetics, 24(10), 2966-2984 (2015-02-06)
We recently identified a novel susceptibility variant, rs865686, for estrogen-receptor positive breast cancer at 9q31.2. Here, we report a fine-mapping analysis of the 9q31.2 susceptibility locus using 43 160 cases and 42 600 controls of European ancestry ascertained from 52
Wesley C Warren et al.
Genome research, 25(12), 1921-1933 (2015-09-18)
We describe a genome reference of the African green monkey or vervet (Chlorocebus aethiops). This member of the Old World monkey (OWM) superfamily is uniquely valuable for genetic investigations of simian immunodeficiency virus (SIV), for which it is the most

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service