Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V003630

Sigma-Aldrich

Xanthan Gum

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
Pricing and availability is not currently available.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Becker et al.
Applied microbiology and biotechnology, 50(2), 145-152 (1998-10-09)
Xanthan gum is a complex exopolysaccharide produced by the plant-pathogenic bacterium Xanthomonas campestris pv. campestris. It consists of D-glucosyl, D-mannosyl, and D-glucuronyl acid residues in a molar ratio of 2:2:1 and variable proportions of O-acetyl and pyruvyl residues. Because of
Hemant R Badwaik et al.
Current drug delivery, 10(5), 587-600 (2013-04-24)
Xanthan gum is a high molecular weight natural polysaccharide produced by fermentation process. It consists of 1, 4-linked β-D-glucose residues, having a trisaccharide side chain attached to alternate D-glucosyl residues. Although the gum has many properties desirable for drug delivery
Hyun M Cho et al.
Journal of the Academy of Nutrition and Dietetics, 115(1), 106-111 (2014-12-03)
Cold beverages are commonly thickened with commercial gum-based food thickeners for consumption by patients with dysphagia. In this study, the rheological properties of a thickened water and five thickened beverages (orange juice, apple juice, grape juice, whole milk, and a
Rebecca J Leonard et al.
Journal of the Academy of Nutrition and Dietetics, 114(4), 590-594 (2013-10-29)
Bolus manipulation is a primary treatment strategy in the management of oral-pharyngeal dysphagia. The use of thickening agents to alter bolus rheology is particularly commonplace; however, the precise effects of these alterations on swallowing remain uncertain. The purpose of our

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service