Skip to Content
MilliporeSigma
All Photos(1)

Documents

V001383

Sigma-Aldrich

Aluminum fluoride

97%

Synonym(s):

Aluminum trifluoride, Trifluoroaluminum

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
AlF3
CAS Number:
Molecular Weight:
83.98
EC Number:
MDL number:
PubChem Substance ID:

Assay

97%

mp

1290 °C (lit.)

density

3.1 g/mL at 25 °C (lit.)

SMILES string

F[Al](F)F

InChI

1S/Al.3FH/h;3*1H/q+3;;;/p-3

InChI key

KLZUFWVZNOTSEM-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Wittinghofer
Current biology : CB, 7(11), R682-R685 (1998-02-28)
Recent three-dimensional structures of phosphoryl transfer enzymes in their aluminum fluoride bound state and corresponding biochemical data have shown how diverse biological problems can be investigated using this small inorganic molecule.
Kristell L S Chatalic et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 55(12), 2050-2056 (2014-11-22)
Gastrin-releasing peptide receptor (GRPR) is overexpressed in human prostate cancer and is being used as a target for molecular imaging. In this study, we report on the direct comparison of 3 novel GRPR-targeted radiolabeled tracers: Al(18)F-JMV5132, (68)Ga-JMV5132, and (68)Ga-JMV4168 (JMV5132
Kan Usuda et al.
Biological trace element research, 167(1), 84-90 (2015-03-10)
Various fluoride compounds are widely used in industry. The present risk assessment study was conducted using a series of inorganic binary fluorides of the type XFn, where X(n) = Na(+), K(+), Li(+), Mg(2+), Ca(2+), Sr(2+), Ba(2+), Al(3+), Nd(3+), La(3+), Ce(3+), Sm(3+), Gd(3+)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service