Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V001204

Sigma-Aldrich

Saccharin

puriss., 98%

Synonym(s):

2,3-Dihydroxy-1,2-benzisothiazol-3-one-1,1-dioxide, 2-Sulfobenzoic acid imide, o-Benzoic sulfimide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5NO3S
CAS Number:
Molecular Weight:
183.18
Beilstein:
6888
EC Number:
MDL number:
PubChem Substance ID:

grade

puriss.

Quality Level

Assay

98%

mp

226-229 °C (lit.)

SMILES string

O=C1NS(=O)(=O)c2ccccc12

InChI

1S/C7H5NO3S/c9-7-5-3-1-2-4-6(5)12(10,11)8-7/h1-4H,(H,8,9)

InChI key

CVHZOJJKTDOEJC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

A sweet tastant for mammals. A glycerol taste receptor binding site specific for glucose has been proposed in drosophila.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M D Reuber
Environmental health perspectives, 25, 173-200 (1978-08-01)
Saccharin is carcinogenic for the urinary bladder in rats and mice, and most likely is carcinogenic in human beings. The neoplasms of the urinary bladder are malignant and invade and metastasize. Male rats are more susceptible to urinary bladder carcinogenesis
J Whysner et al.
Pharmacology & therapeutics, 71(1-2), 225-252 (1996-01-01)
Sodium saccharin (NaSac) produces bladder tumors consistently in male rats only after lifetime exposure that begins at birth. NaSac is not metabolized and is negative in most genotoxicity tests. NaSac-induced cell damage and proliferation have been proposed as important factors
R L Anderson
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 26(7), 637-644 (1988-07-01)
An hypothesis is presented of a mechanism for the sodium saccharin (NaS)-associated tumorigenesis of the urinary bladder that occurs in male rats. The ingestion of high doses of NaS is associated with increased urine volume and bladder mass. In rats
D L Arnold et al.
Toxicology, 27(3-4), 179-256 (1983-07-01)
Saccharin, first synthesized in 1879, eventually became popular as an inexpensive substitute for sugar, particularly as a non-caloric sweetner. The dispute concerning the safety of saccharin for human consumption is almost as old as saccharin itself. In this article, the
Marilyn E Carroll et al.
Behavioural pharmacology, 19(5-6), 435-460 (2008-08-12)
A positive relationship between the consumption of sweetened dietary substances (e.g. saccharin and sucrose) and drug abuse has been reported in both the human and other animal literature. The proposed genetic contribution to this relationship has been based on evidence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service