Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V000954

Sigma-Aldrich

Sodium rhodizonate dibasic

p.a., 98.5%

Synonym(s):

3,4,5,6-Tetraoxocyclohexene-1,2-diol disodium salt, Rhodizonic acid disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6Na2O6
CAS Number:
Molecular Weight:
214.04
Beilstein:
5181379
EC Number:
MDL number:
PubChem Substance ID:

grade

p.a.

Quality Level

Assay

98.5%

mp

>300 °C (lit.)

SMILES string

[Na+].[Na+].[O-]C1=C([O-])C(=O)C(=O)C(=O)C1=O

InChI

1S/C6H2O6.2Na/c7-1-2(8)4(10)6(12)5(11)3(1)9;;/h7-8H;;/q;2*+1/p-2

InChI key

SXWPCWBFJXDMAI-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service