Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V000538

Sigma-Aldrich

Copper(II) carbonate basic

basic, 52-56%

Synonym(s):

Cupric carbonate basic

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CuCO3 · Cu(OH)2
CAS Number:
Molecular Weight:
221.12
EC Number:
MDL number:
PubChem Substance ID:

concentration

52-56%

SMILES string

[Cu++].O[Cu]O.[O-]C([O-])=O

InChI

1S/CH2O3.2Cu.2H2O/c2-1(3)4;;;;/h(H2,2,3,4);;;2*1H2/q;2*+2;;/p-4

InChI key

ZMMDPCMYTCRWFF-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service