Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I10006

Dichloromethane

suitable for HPLC, ≥99.7%

Synonym(s):

Methylene chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH2Cl2
CAS Number:
Molecular Weight:
84.93
Beilstein:
1730800
EC Number:
MDL number:
UNSPSC Code:
41116004
PubChem Substance ID:
Assay:
≥99.7%
technique(s):
HPLC: suitable
bp:
39.8-40 °C (lit.)
vapor pressure:
24.45 psi ( 55 °C)
6.83 psi ( 20 °C)
Pricing and availability is not currently available.

vapor density

2.9 (vs air)

Quality Level

vapor pressure

24.45 psi ( 55 °C)
6.83 psi ( 20 °C)

product line

Vetec

Assay

≥99.7%

form

liquid

autoignition temp.

1223 °F

expl. lim.

22 %

technique(s)

HPLC: suitable

refractive index

n20/D 1.424 (lit.)

bp

39.8-40 °C (lit.)

mp

−95 °C (lit.)

density

1.325 g/mL at 25 °C (lit.)

SMILES string

ClCCl

InChI

1S/CH2Cl2/c2-1-3/h1H2

InChI key

YMWUJEATGCHHMB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aria Nouri et al.
Spine, 40(14), 1092-1100 (2015-04-22)
Ambispective study. To determine whether MRI parameters improve the predictive performance of a validated clinical prediction rule used to assess functional outcomes in surgical patients with DCM. Degenerative cervical myelopathy (DCM) is the most common cause of spinal cord dysfunction
Damian M Herz et al.
Brain : a journal of neurology, 138(Pt 6), 1658-1666 (2015-04-18)
Dopaminergic signalling in the striatum contributes to reinforcement of actions and motivational enhancement of motor vigour. Parkinson's disease leads to progressive dopaminergic denervation of the striatum, impairing the function of cortico-basal ganglia networks. While levodopa therapy alleviates basal ganglia dysfunction
Molamma P Prabhakaran et al.
Materials science & engineering. C, Materials for biological applications, 56, 66-73 (2015-08-08)
Advanced engineering of materials for the development of drug delivery devices provides scope for novel and versatile strategies for treatment of various diseases. 'Electrospraying' was used to prepare PLGA microparticles and further encapsulate the drug, metronidazole (Met) within the particles
Angharad M Roberts et al.
Science translational medicine, 7(270), 270ra6-270ra6 (2015-01-16)
The recent discovery of heterozygous human mutations that truncate full-length titin (TTN, an abundant structural, sensory, and signaling filament in muscle) as a common cause of end-stage dilated cardiomyopathy (DCM) promises new prospects for improving heart failure management. However, realization
Ludivine Renaud et al.
Circulation. Heart failure, 8(6), 1094-1104 (2015-09-16)
MicroRNAs (miRNAs) and histone deacetylases (HDACs) serve a significant role in the pathogenesis of a variety of cardiovascular diseases. The transcriptional regulation of miRNAs is poorly understood in cardiac hypertrophy. We investigated whether the expression of miR-133a is epigenetically regulated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service