Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1724918

USP

Zolpidem Related Compound A

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

N,N-Dimethyl-2-[7-methyl-2-(4-methylphenyl)imidazo[1,2-a]pyridin-3-yl]acetamide

Sign Into View Organizational & Contract Pricing

Select a Size

15 MG
$1,850.00

$1,850.00


Please contact Customer Service for Availability


Select a Size

Change View
15 MG
$1,850.00

About This Item

Empirical Formula (Hill Notation):
C19H21N3O
Molecular Weight:
307.39
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$1,850.00


Please contact Customer Service for Availability

API family

zolpidem

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

SMILES string

CC1=CC=C(C=C1)C2=C(CC(N(C)C)=O)N3C=CC(C)=CC3=N2

InChI

1S/C19H21N3O/c1-13-5-7-15(8-6-13)19-16(12-18(23)21(3)4)22-10-9-14(2)11-17(22)20-19/h5-11H,12H2,1-4H3

InChI key

CZVNHSKGTUTQHR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Zolpidem Related Compound A USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Zolpidem Tartrate
  • Zolpidem Tartrate Extended-Release Tablets
  • Zolpidem Tartrate Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

USP issued SDS can be found here.
Sales restrictions may apply.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service