Skip to Content
MilliporeSigma
All Photos(1)

Documents

1699333

USP

Trospium chloride Related Compound C

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(1R,3r,5S)-3-Hydroxyspiro[8-azoniabicyclo[3.2.1]octane-8,1′-pyrrolidinium] chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H20ClNO
CAS Number:
Molecular Weight:
217.74
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

trospium

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

InChI

1S/C11H20NO.ClH/c13-11-7-9-3-4-10(8-11)12(9)5-1-2-6-12;/h9-11,13H,1-8H2;1H/q+1;/p-1/t9-,10+,11+;

InChI key

HZDYZXDBRXHZIX-JGNDIBIWSA-M

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Trospium chloride Related Compound C USP reference standard for use in specified quality tests and assays.

Also used to prepare standard solution during the impurity analysis of trospium chloride by using liquid chromatography in conjunction with a refractive index (RI) detector.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Trospium Chloride
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 38(6), 4541-4541 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service