Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1672698

USP

Trandolapril Related Compound A

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(2S,3aR,7aS)-1-[(2S)-2-[[(1S)-1-(Methoxycarbonyl)-3-phenylpropyl]amino]-1-oxopropyl]octahydro-1H-indole-2-carboxylic acid, Trandolapril methyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
A$1,040.00

A$1,040.00


Availability

Estimated to ship onMarch 10, 2025


Request a Bulk Order

Select a Size

Change View
100 μL
A$1,040.00

About This Item

Empirical Formula (Hill Notation):
C23H32N2O5
CAS Number:
Molecular Weight:
416.51
UNSPSC Code:
41116107
NACRES:
NA.24

A$1,040.00


Availability

Estimated to ship onMarch 10, 2025


Request a Bulk Order

grade

pharmaceutical primary standard

API family

trandolapril

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

InChI

1S/C23H32N2O5/c1-15(24-18(23(29)30-2)13-12-16-8-4-3-5-9-16)21(26)25-19-11-7-6-10-17(19)14-20(25)22(27)28/h3-5,8-9,15,17-20,24H,6-7,10-14H2,1-2H3,(H,27,28)/t15-,17+,18-,19-,20-/m0/s1

InChI key

QUDOFHJAZMIEDX-ZDNSCYFHSA-N

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
HPA047860HPA056304SAB4500656
Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

product line

Prestige Antibodies® Powered by Atlas Antibodies

product line

Prestige Antibodies® Powered by Atlas Antibodies

product line

Prestige Antibodies® Powered by Atlas Antibodies

product line

-

Gene Information

human ... SEPTIN10(151011)

Gene Information

human ... SEPTIN10(151011)

Gene Information

human ... SEPT10(151011)

Gene Information

human ... SEPT1(1731)

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Trandolapril Related Compound A USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monograph such as Trandolapril and Verapamil Hydrochloride Extended-Release Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


  • Certificates of Analysis (COA)

    Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

    Need A Sample COA?

    This is a sample Certificate of Analysis (COA) and may not represent a recently manufactured lot of this specific product.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service