Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1651654

USP

2,3,5,4′-Tetrahydroxystilbene-2-O-Beta-D-Glucoside

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

2,3,5,4′-Tetrahydroxystilbene 2-O-β-D-glucoside, (E)-2,4-dihydroxy-6-(4-hydroxystyryl)phenyl β-D-glucopyranoside, 2,4-Dihydroxy-6-[(1E)-2-(4-hydroxyphenyl)ethenyl]phenyl β-D-Glucopyranoside, EH-201, THSG, Tetrahydroxystilbene glucoside

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$364.00

$364.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
$364.00

About This Item

Empirical Formula (Hill Notation):
C20H22O9
CAS Number:
Molecular Weight:
406.38
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$364.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

manufacturer/tradename

USP

application(s)

pharmaceutical

format

neat

storage temp.

2-8°C

SMILES string

O1[C@H]([C@@H]([C@H]([C@@H]([C@H]1CO)O)O)O)Oc2c(cc(cc2\C=C\c3ccc(cc3)O)O)O

InChI

1S/C20H22O9/c21-9-15-16(25)17(26)18(27)20(28-15)29-19-11(7-13(23)8-14(19)24)4-1-10-2-5-12(22)6-3-10/h1-8,15-18,20-27H,9H2/b4-1+/t15-,16-,17+,18-,20+/m1/s1

InChI key

JAYVHSBYKLLDJC-DSNJPTTOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Biochem/physiol Actions

2,3,5,4′-Tetrahydroxystilbene 2-O-β-D-glucoside induces the expression and enhances the activity of EPO.
2,3,5,4′-Tetrahydroxystilbene 2-O-β-D-glucoside induces the expression and enhances the activity of EPO. The compound stimulates mitochondrial biogenesis, and expression of hemoglobin in non-hematopoietic cells. 2,3,5,4′-Tetrahydroxystilbene 2-O-β-D-glucoside has demonstrated protective effects in cardiovascular disease and ischemia models.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.
USP issued SDS can be found here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service