Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1467837

USP

PS NMR System Suitability

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Hyaluronate sodium oligomer, Hyaluronic acid oligosaccharide

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$837.00

$837.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$837.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$837.00


Please contact Customer Service for Availability

Request a Bulk Order

packaging

pkg of 168 mg

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia

Application

PS NM System Suitability reference standard for use in specified quality tests and assays.

Also used to prepare system suitability solutions according to the United States Pharmacopeia (USP) general chapter. [1]

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

This product is part of the USP Biologics program.
Sales restrictions may apply.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

{?198? Nuclear magnetic resonance spectroscopy identity testing of bacterial polysaccharides used in vaccine manufacture}
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 43(2), 6596-6596 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service