Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1424062

USP

Methyl acrylate, Methyl methacrylate and Methycrylic acid (7:3:1) Copolymer 280000 Dispersion

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Poly(methyl acrylate-co-methyl methacrylate-co-methacrylic acid) 7:3:1

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24

packaging

pkg of 200 mg

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Methyl acrylate, Methyl methacrylate and Methycrylic acid (7:3:1) Copolymer 280000 Dispersion USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.
Also, for use with USP monograph such as Methyl Acrylate, Methyl Methacrylate and Methacrylic Acid (7 to 3 to 1) Copolymer 280000 Dispersion

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

Sales restrictions may apply.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service