Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1366578

USP

Linalyl acetate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

3,7-Dimethyl-1,6-octadien-3-yl acetate, Bergamol

Sign Into View Organizational & Contract Pricing

Select a Size

0.3 ML
$681.00

$681.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
0.3 ML
$681.00

About This Item

Linear Formula:
CH3CO2C(CH=CH2)(CH3)CH2CH2CH=C(CH3)2
CAS Number:
Molecular Weight:
196.29
Beilstein:
1724500
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$681.00


Please contact Customer Service for Availability

Request a Bulk Order

vapor density

6.8 (vs air)

vapor pressure

0.1 mmHg ( 20 °C)

API family

linalyl acetate

packaging

pkg of 0.3 mL

manufacturer/tradename

USP

refractive index

n20/D 1.453 (lit.)

bp

220 °C (lit.)

density

0.901 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

C\C(C)=C\CCC(C)(OC(C)=O)C=C

InChI

1S/C12H20O2/c1-6-12(5,14-11(4)13)9-7-8-10(2)3/h6,8H,1,7,9H2,2-5H3

InChI key

UWKAYLJWKGQEPM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Linalyl acetate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service