Skip to Content
MilliporeSigma

Skip To

1362500

USP

Levonordefrin

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(−)-3,4-Dihydroxynorephedrine, α-(1-Aminoethyl)-3,4-dihydroxybenzyl alcohol, (−)-α-Methylnorepinephrine, 4-(2-Amino-1-hydroxypropyl)-1,2-benzenediol, L-4-(2-Amino-1-hydroxypropyl)-1,2-benzenediol, Levonordefrin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
200 mg
Please contact Customer Service for Availability
$579.00

About This Item

Linear Formula:
(HO)2C6H3CH(OH)CH(CH3)NH2
CAS Number:
Molecular Weight:
183.20
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:

$579.00


Please contact Customer Service for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

levonordefrin

manufacturer/tradename

USP

application(s)

pharmaceutical
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

C[C@H](N)[C@H](O)c1ccc(O)c(O)c1

InChI

1S/C9H13NO3/c1-5(10)9(13)6-2-3-7(11)8(12)4-6/h2-5,9,11-13H,10H2,1H3/t5-,9-/m0/s1

InChI key

GEFQWZLICWMTKF-CDUCUWFYSA-N

Gene Information

General description

Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Levonordefrin USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Mepivacaine Hydrochloride and Levonordefrin Injection

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML0675SMB00193
description

United States Pharmacopeia (USP) Reference Standard

description

≥95% (HPLC)

description

≥95% (LC/MS-ELSD)

grade

pharmaceutical primary standard

grade

-

grade

-

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

format

neat

format

-

format

-

application(s)

pharmaceutical (small molecule)

application(s)

-

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

Gene Information

human ... ADRA2A(150), ADRA2B(151), ADRA2C(152)

Gene Information

-

Gene Information

-

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters