Skip to Content
MilliporeSigma
All Photos(1)

Documents

1340009

USP

Indocyanine green

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Cardiogreen, Foxgreen, IC Green, Indocyanine green

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H47N2NaO6S2
CAS Number:
Molecular Weight:
774.96
Beilstein:
4115884
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

indocyanine green

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[Na+].CC1(C)/C(=C\C=C\C=C\C=C\C2=[N+](CCCCS([O-])(=O)=O)c3ccc4ccccc4c3C2(C)C)N(CCCCS([O-])(=O)=O)c5ccc6ccccc6c15

InChI

1S/C43H48N2O6S2.Na/c1-42(2)38(44(28-14-16-30-52(46,47)48)36-26-24-32-18-10-12-20-34(32)40(36)42)22-8-6-5-7-9-23-39-43(3,4)41-35-21-13-11-19-33(35)25-27-37(41)45(39)29-15-17-31-53(49,50)51;/h5-13,18-27H,14-17,28-31H2,1-4H3,(H-,46,47,48,49,50,51);/q;+1/p-1

InChI key

MOFVSTNWEDAEEK-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application


  • Evaluation of gastric tube blood flow by multispectral camera and fluorescence angiography: This study demonstrates the application of Indocyanine green in assessing the blood flow in gastric tubes using multispectral imaging techniques, contributing to enhanced surgical outcomes and patient safety in gastrointestinal operations (Haruta et al., 2024).


Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Indocyanine Green
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 47(2), 2322-2322 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service