Skip to Content
MilliporeSigma
All Photos(1)

Documents

1336001

USP

Idoxuridine

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

5-Iodo-2′-deoxyuridine, 1-(2-Deoxy-β-D-ribofuranosyl)-5-iodouracil, 2′-Deoxy-5-iodouridine, 5-IUdR, IDU, IdUrd, Idoxuridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11IN2O5
CAS Number:
Molecular Weight:
354.10
Beilstein:
30397
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

idoxuridine

manufacturer/tradename

USP

mp

194 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

OC[C@H]1O[C@H](C[C@@H]1O)N2C=C(I)C(=O)NC2=O

InChI

1S/C9H11IN2O5/c10-4-2-12(9(16)11-8(4)15)7-1-5(14)6(3-13)17-7/h2,5-7,13-14H,1,3H2,(H,11,15,16)/t5-,6+,7+/m0/s1

InChI key

XQFRJNBWHJMXHO-RRKCRQDMSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Idoxuridine USP Reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monograph such as Idoxuridine Ophthalmic Solution

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Muta. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service