Skip to Content
MilliporeSigma
All Photos(1)

Documents

1254519

USP

Estriol Related Compound A

United States Pharmacopeia (USP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

estriol

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application


  • Robust Strategies to Eliminate Endocrine Disruptive Estrogens: Highlighting the significance of estriol related compounds, this study explores advanced strategies to eliminate endocrine disruptive estrogens from water resources. Such research is crucial for ensuring the safety and quality of environmental water sources, with implications for public health and ecosystem stability (Bilal et al., 2022).

  • Effectiveness of Vaginal Hyaluronic Acid versus Estriol: Addressing the comparative effectiveness of vaginal treatments, this research evaluates hyaluronic acid against estriol related compounds for relieving vaginal dryness. The study contributes to gynecological healthcare by providing evidence-based insights into effective treatments (Stute, 2013).

  • Environmental Impact of Estrogens: This environmental toxicology research focuses on the analytical methods for detecting endocrine-disrupting chemicals, including estriol related compounds, in various environmental matrices. Such studies are essential for understanding and mitigating the environmental impacts of synthetic estrogens, which pose risks to wildlife and human health (Ikonomou et al., 2008).

  • Advanced Detection of Estrogens: Innovating in the field of chromatography, this study develops methods for the detection of estrogens, including estriol related compounds, in environmental samples. This work enhances our capabilities in monitoring and managing the presence of hormone disruptors in the ecosystem, providing crucial data for environmental protection and regulatory compliance (López de Alda & Barceló, 2001).

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service