Skip to Content
MilliporeSigma
All Photos(1)

Documents

1076035

USP

[des-Glu13]-Bivalirudin

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

D-Phenylalanyl-L-prolyl-L-arginyl-L-prolylglycylglycylglycylglycyl-L-asparaginylglycyl -L-alpha-aspartyl-L-phenylalanyl-L-alpha-glutamyl-L-isoleucyl-L-prolyl-L-alpha-glutamyl-L-alpha-glutamyl-L-tyrosyl-L-leucine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C93H131N23O30
CAS Number:
Molecular Weight:
2051.17
UNSPSC Code:
41116107
NACRES:
NA.24

API family

bivalirudin

packaging

pkg of 1 mg

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

[des-Glu13]-Bivalirudin USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monograph such as Bivalirudin.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.

Other Notes

This product is part of the USP Biologics program.
Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service