Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z777279

Antibodix® WCX-NP (1.7 μm) Weak Cation Exchange HPLC Columns

L × I.D. 1 cm × 4 mm, HPLC Guard Cartridge and Holder

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$1,020.00

$1,020.00


Please contact Customer Service for Availability


Select a Size

Change View
1 PKG
$1,020.00

About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

$1,020.00


Please contact Customer Service for Availability

Product Name

Antibodix® WCX-NP1.7 Weak Cation Exchange Guard Cartridge and Holder, and Holder, L × I.D. 1 cm × 4 mm, 1.7 μm particle size

material

stainless steel

product line

Antibodix®

manufacturer/tradename

Sepax 602NP2-4001C

packaging

1 ea of

parameter

69 Mpa max. pressure (10000 psi)
80 °C max. temp.

technique(s)

HPLC: suitable

L × I.D.

1 cm × 4 mm

matrix

polymeric

matrix active group

weak cation exchange phase

particle size

1.7 μm

pore size

non-porous

operating pH range

2.0-12.0

separation technique

cation exchange

Looking for similar products? Visit Product Comparison Guide

General description

The non-porous poly(styrene divinylbenzene) particles in Antibodix columns are shielded by a hydrophilic polymer layer that contains a network of weak (WCX) cation exchange groups. Antibodix columns are designed for the reproducible analysis of charge variants of monoclonal antibodies by salt or pH gradient.

Antibodix User Manual

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Antibodix is a registered trademark of Sepax Technologies

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service