Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z777249

Proteomix® WAX-NP3 (3 µm) HPLC Columns

L × I.D. 1 cm × 2 mm

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$921.00

$921.00


Please contact Customer Service for Availability


Select a Size

Change View
1 PKG
$921.00

About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

$921.00


Please contact Customer Service for Availability

Product Name

Proteomix® WAX-NP3 Weak Anion Exchange Guard Cartridge and Holder, L × I.D. 1 cm × 2 mm, 3 μm particle size

material

stainless steel

product line

Proteomix®

manufacturer/tradename

Sepax 404NP3-2001C

packaging

1 ea of

parameter

55 Mpa max. pressure (8000 psi)
80 °C max. temp.

technique(s)

HPLC: suitable

L × I.D.

1 cm × 2 mm

matrix

spherical PS-DVB
polymeric

matrix active group

weak anion exchange phase

particle size

3 μm

pore size

non-porous

operating pH

2.0-12.0

separation technique

anion exchange

Looking for similar products? Visit Product Comparison Guide

General description

Proteomix columns are packed with poly(styrene divinylbenzene) particles that are protected with a hydrophilic polymer layer that contains a network of weak (WCX) or strong (SCX) cation exchange groups. Use Proteomix cation-exchange columns for the analysis or small scale purification of positively charged biopolymers including glycoprotein isoforms and mAb variants.

Proteomix User Manual

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Proteomix is a registered trademark of Sepax Technologies

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service