Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z226963

Si Guard Cartridge

5 μm particle size, L × I.D. 1 cm × 4.6 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

Product Name

Spherisorb® Si Guard Cartridge, 5 μm particle size, L × I.D. 1 cm × 4.6 mm

Agency

suitable for USP L3

product line

Spherisorb®

packaging

pkg of 4 ea

technique(s)

HPLC: suitable

L × I.D.

1 cm × 4.6 mm

matrix active group

silica phase

particle size

5 μm

compatibility

for use with 3.0 to 4.6 mm column

separation technique

hydrophilic interaction (HILIC)
normal phase

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Spherisorb is a registered trademark of Waters

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service