Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z226106

Spherisorb® Phenyl HPLC Column

5 μm particle size, L × I.D. 25 cm × 4.6 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

feature

endcapped: no

manufacturer/tradename

Spherisorb®

extent of labeling

2.5% Carbon loading

parameter

20-45 °C temp. range
400 bar pressure (5801 psi)

technique(s)

HPLC: suitable

L × I.D.

25 cm × 4.6 mm

surface area

220 m2/g

matrix

phenyl

matrix active group

phenyl phase

particle size

5 μm

pore size

80 Å

separation technique

normal phase

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Spherisorb is a registered trademark of Waters

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service