Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Y0002213

DL-Lysine acetylsalicylate impurity G

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

N2,N6-Diacetyllysine

Sign Into View Organizational & Contract Pricing

Select a Size

15 MG
$169.00

$169.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
15 MG
$169.00

About This Item

Empirical Formula (Hill Notation):
C10H18N2O4
CAS Number:
Molecular Weight:
230.26
UNSPSC Code:
41116107
NACRES:
NA.24

$169.00


Please contact Customer Service for Availability

Request a Bulk Order

API family

lysine

manufacturer/tradename

EDQM

storage temp.

2-8°C

SMILES string

N(C(CCCCNC(=O)C)C(=O)O)C(=O)C

InChI

1S/C10H18N2O4/c1-7(13)11-6-4-3-5-9(10(15)16)12-8(2)14/h9H,3-6H2,1-2H3,(H,11,13)(H,12,14)(H,15,16)

InChI key

ZHZUEHHBTYJTKY-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

ᴅʟ-Lysine acetylsalicylate impurity G EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service