Y0002167
Rosuvastatin impurity mixture
CRS, European Pharmacopoeia (EP) Reference Standard
Select a Size
About This Item
Recommended Products
API family
rosuvastatin
manufacturer/tradename
EDQM
application(s)
pharmaceutical
storage temp.
2-8°C
General description
Application
Packaging
Other Notes
related product
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service