SU860006
Inter-Seal™ bonded caps (inseparable caps and liner)
white polypropylene cap (solid), tan PTFE/silicone (EPA Quality), thickness 3.2 mm, thread for 24-400, for use with 20 or 40mL vial, pkg of 100 ea
About This Item
Recommended Products
material
tan PTFE/silicone (EPA Quality)
white polypropylene cap (solid)
feature
InterSeal™ (cap and liner form an unseparatable unit)
hardness
45 (Durometer: shore A)
packaging
pkg of 100 ea
thickness
3.2 mm
fitting
thread for 24-400
compatibility
for use with 20 or 40mL vial
Related Categories
General description
Benefits include:
- Significant bond strength
- Withstands multiple injections
- Resistant to coring
- Excellent resealability
- Broad chemical resistance
Legal Information
related product
Choose from one of the most recent versions:
Certificates of Analysis (COA)