Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAAR6725000

Water

Deionized, UniLAB

Synonym(s):

Water

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2O
CAS Number:
Molecular Weight:
18.02
Beilstein:
2050024
MDL number:
UNSPSC Code:
12191602

product name

Water de-ionised, uniLAB

product line

uniLAB

packaging

poly bottle of 4 × 5 L (SAAR6725000LL)

refractive index

n20/D 1.34 (lit.)

bp

100 °C (lit.)

mp

0 °C (lit.)

density

1.000 g/mL at 3.98 °C (lit.)

storage temp.

2-30°C

SMILES string

O

InChI

1S/H2O/h1H2

InChI key

XLYOFNOQVPJJNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chemically pure
For laboratory use only

Legal Information

uniLAB is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service