Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAAR1765000

Cyclohexanone

uniLAB

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H10(=O)
CAS Number:
Molecular Weight:
98.14
Beilstein:
385735
MDL number:
UNSPSC Code:
12191502

vapor density

3.4 (vs air)

vapor pressure

3.4 mmHg ( 20 °C)

product line

uniLAB

autoignition temp.

788 °F

expl. lim.

1.1 %, 100 °F
9.4 %

packaging

glass bottle of 4 × 2.5 L (SAAR1765000LC)

refractive index

n20/D 1.450 (lit.)

bp

155 °C (lit.)

mp

−47 °C (lit.)

density

0.947 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

O=C1CCCCC1

InChI

1S/C6H10O/c7-6-4-2-1-3-5-6/h1-5H2

InChI key

JHIVVAPYMSGYDF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chemically pure
For laboratory use only

Legal Information

uniLAB is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

111.2 °F - closed cup

Flash Point(C)

44 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service