Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAAR1202000

Auramine O

Synonym(s):

4,4′-(Imidocarbonyl)bis(N,N-dimethylaniline) monohydrochloride, Basic Yellow 2, Pyoctaninum aureum

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H21N3 · HCl
CAS Number:
Molecular Weight:
303.83
Colour Index Number:
41000
Beilstein:
4030061
MDL number:
UNSPSC Code:
12352116

packaging

pkg of 100 g (SAAR1202000DC)

mp

>250 °C (dec.) (lit.)

storage temp.

2-30°C

SMILES string

Cl[H].CN(C)c1ccc(cc1)C(=N)c2ccc(cc2)N(C)C

InChI

1S/C17H21N3.ClH/c1-19(2)15-9-5-13(6-10-15)17(18)14-7-11-16(12-8-14)20(3)4;/h5-12,18H,1-4H3;1H

InChI key

KSCQDDRPFHTIRL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

For laboratory use only

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service