Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAAR1122720

Ammonium chloride

univAR

Synonym(s):

Salmiac

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4Cl
CAS Number:
Molecular Weight:
53.49
Beilstein:
4371014
MDL number:
UNSPSC Code:
12352302

vapor density

1.9 (vs air)

vapor pressure

1 mmHg ( 160.4 °C)

product line

univAR

packaging

pkg of 20 kg (SAAR1122720GL)
poly bottle of 12 × 500 g (SAAR1122720EM)

mp

340 °C (subl.) (lit.)

storage temp.

2-30°C

SMILES string

N.Cl

InChI

1S/ClH.H3N/h1H;1H3

InChI key

NLXLAEXVIDQMFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Analysed reagent
For laboratory use only

Legal Information

univAR is a trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service