Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PS901

Dimethyl phthalate

analytical standard

Synonym(s):

DMP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,2-(CO2CH3)2
CAS Number:
Molecular Weight:
194.18
Beilstein:
1911460
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

autoignition temp.

1033 °F

packaging

ampule of 1000 mg

manufacturer/tradename

Chem Service, Inc. PS-901

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.515 (lit.)

bp

282 °C (lit.)

mp

2 °C (lit.)

density

1.19 g/mL at 25 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

COC(=O)c1ccccc1C(=O)OC

InChI

1S/C10H10O4/c1-13-9(11)7-5-3-4-6-8(7)10(12)14-2/h3-6H,1-2H3

InChI key

NIQCNGHVCWTJSM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

294.8 °F - closed cup

Flash Point(C)

146.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenxin Wang et al.
Environmental monitoring and assessment, 184(11), 6789-6798 (2011-12-08)
Atmospheric trace phthalic esters (PAEs) carried by total suspended particulates were systematically investigated. A total of 450 air samples were collected at six typical locations and three specific micro-environmental sites (a stadium with a new plastic track, new cars, and
L J Xu et al.
Ultrasonics sonochemistry, 20(3), 892-899 (2012-12-05)
A comprehensive study of the sonochemical degradation of dimethyl phthalate (DMP) was carried out using high-frequency ultrasonic processes. The effects of various operating parameters were investigated, including ultrasonic frequency, power density, initial DMP concentration, solution pH and the presence of
L J Xu et al.
Water research, 47(6), 1996-2004 (2013-02-12)
The merits of the combined process of high-frequency ultrasound (US) and catalyst-free ultraviolet irradiation (UV) have been evaluated in this study by investigating the sonophotolytic degradation of dimethyl phthalate (DMP). A 400 kHz ultrasonic system and a photolytic system at
Yiling Chen et al.
Journal of hazardous materials, 235-236, 92-100 (2012-08-14)
In this study, we demonstrate that the decomposition of dimethyl phthalate under microwave irradiation could be greatly enhanced over Fe@Fe(2)O(3) nanowires supported on activated carbon (Fe@Fe(2)O(3)/AC). The great enhanced decomposition of dimethyl phthalate could be attributed to a unique microwave
Zhengwen Xu et al.
Environmental science & technology, 44(8), 3130-3135 (2010-03-17)
In the present study, a novel approach was developed to remove dimethyl phthalate (DMP), a representative phthalic acid ester (PAE) pollutant, from an aqueous solution using a macroporous OH-type strong base anion exchange resin D201-OH. As compared to the traditional

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service