Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PS609

Monocrotophos

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14NO5P
CAS Number:
Molecular Weight:
223.16
EC Number:
MDL number:
UNSPSC Code:
41116107
Pricing and availability is not currently available.

grade

analytical standard

packaging

ampule of 250 mg

manufacturer/tradename

Chem Service, Inc. PS-609

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

CNC(=O)\C=C(/C)OP(=O)(OC)OC

InChI

1S/C7H14NO5P/c1-6(5-7(9)8-2)13-14(10,11-3)12-4/h5H,1-4H3,(H,8,9)/b6-5+

InChI key

KRTSDMXIXPKRQR-AATRIKPKSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Muta. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W Mücke
Reviews of environmental contamination and toxicology, 139, 59-65 (1994-01-01)
Regarding the metabolic fate of monocrotophos, it appears that the mechanisms summarized below are largely independent of the animal species. Furthermore, significant sex dependency of any of these parameters has not been found. ABSORPTION: After oral administration to rats and
Amajad Iqbal Kazi et al.
Neurotoxicology, 33(2), 156-161 (2012-01-31)
Neurotoxicity of organophosphate pesticide poisoning, a lead cause of death in South Asia, has not been clearly elucidated. Organophosphates inhibit acetylcholinesterase and neurotoxicity is primarily a result of acetylcholine induced hyperactivation in different regions of the brain. Neurotoxicity also results
T Skripsky et al.
Reviews of environmental contamination and toxicology, 139, 13-39 (1994-01-01)
Monocrotophos is a water-soluble organophosphate insecticide with high oral and moderate dermal toxicity. The toxicologically relevant mode of action is the inhibition of ChE activities. The toxicity of organophosphate metabolites of monocrotophos is comparable with the parent compound. Glycol conjugation
E Neuenschwander
Reviews of environmental contamination and toxicology, 139, 41-46 (1994-01-01)
Monocrotophos is an old product that is still very much appreciated as a low-cost and efficient insecticide. Due to its water solubility, formulation recipes are rather simple and no sophisticated equipment is required in a formulation plant. Formulations can easily
Madhusudan Reddy Narra et al.
Toxicology and industrial health, 28(4), 343-352 (2011-10-11)
Sublethal effects of chlorpyrifos (CPF) and monocrotophos (MCP) on fish biochemical constituents were investigated along with the assessment of recovery response after cessation of intoxication. The fish, Clarias batrachus were exposed to 1.656 mg(-l) and 2.114 mg(-l) of CPF and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service