Skip to Content
MilliporeSigma
All Photos(1)

Documents

PS540

Barban

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H9Cl2NO2
CAS Number:
Molecular Weight:
258.10
Beilstein:
2376181
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 100 mg

manufacturer/tradename

Chem Service, Inc. PS-540

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

ClCC#CCOC(=O)Nc1cccc(Cl)c1

InChI

1S/C11H9Cl2NO2/c12-6-1-2-7-16-11(15)14-10-5-3-4-9(13)8-10/h3-5,8H,6-7H2,(H,14,15)

InChI key

MCOQHIWZJUDQIC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jan C Dobrowolski et al.
Journal of chemical information and modeling, 45(4), 1030-1038 (2005-07-28)
The structure and stability of two-component carbyne catenanes, viewed as model compounds for DNA catenanes, have been estimated by molecular mechanics (MM) calculations. The carbyne catenane molecules studied were composed from interwined cyclic molecules constituted solely from carbon atoms bonded
P P Smith et al.
Science (New York, N.Y.), 212(4492), 322-324 (1981-04-17)
High-resolution transmission electron microscopy, shows that carbon in the Allende carbonaceous chondrite meteorite is predominantly a poorly crystalline graphite. Such material is of interest as an important carrier of the isotopically anomalous noble gases found in carbonaceous chondrites.
Carbon allotropes: On the road to carbyne.
Michael M Haley
Nature chemistry, 2(11), 912-913 (2010-10-23)
Z H Xu et al.
Guang pu xue yu guang pu fen xi = Guang pu, 21(5), 596-598 (2003-08-30)
The Raman and infrared spectra of complex: [Cl(CO)2 (DPPE) W identical to CC6H4C identical to CC6H4NC]2 Re Cl(CO)3 (DPPE = bis (diphenylphosphino) ethane, (Ph)2PCH2CH2P(Ph)2) were measured at high external pressure (up to about 50 kbar). The pressure induced phase transition
Synthesis and structural analysis of one-dimensional sp-hybridized carbon chain molecules.
Sanghee Kim
Angewandte Chemie (International ed. in English), 48(42), 7740-7743 (2009-09-12)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service