Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PS2071-250MG

Deltamethrin

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H19Br2NO3
CAS Number:
Molecular Weight:
505.20
Beilstein:
6746312
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 250 mg

manufacturer/tradename

Chem Service, Inc. PS-2071

SMILES string

CC1(C)[C@@H](\C=C(\Br)Br)[C@H]1C(=O)O[C@H](C#N)c2cccc(Oc3ccccc3)c2

InChI

1S/C22H19Br2NO3/c1-22(2)17(12-19(23)24)20(22)21(26)28-18(13-25)14-7-6-10-16(11-14)27-15-8-4-3-5-9-15/h3-12,17-18,20H,1-2H3/t17-,18+,20-/m0/s1

InChI key

OWZREIFADZCYQD-NSHGMRRFSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chen-Chao Shen et al.
Environmental science. Processes & impacts, 17(12), 2074-2081 (2015-10-30)
The aim of the study was to determine whether humic acid (HA) prevented gene and biochemical toxic effects in earthworms (Eisenia foetida) exposed to nickel and deltamethrin (at 100 and 1 mg kg(-1), respectively) in soil. Cellular- and molecular-level toxic
Elvira Zamora Perea et al.
Malaria journal, 8, 208-208 (2009-09-05)
The purpose of this study was to establish whether the "bottle assay", a tool for monitoring insecticide resistance in mosquitoes, can complement and augment the capabilities of the established WHO assay, particularly in resource-poor, logistically challenging environments. Laboratory reared Aedes
Rui-Long Wang et al.
International journal of molecular sciences, 16(9), 22606-22620 (2015-09-24)
Cytochrome P450 monooxygenases (P450s) of insects play crucial roles in the metabolism of endogenous and dietary compounds. Tobacco cutworm moth (Spodoptera litura), an important agricultural pest, causes severe yield losses in many crops. In this study, we identified CYP9A40, a
Sandra M De Urioste-Stone et al.
Transactions of the Royal Society of Tropical Medicine and Hygiene, 109(2), 159-167 (2015-01-22)
Integrated vector management strategies depend on local eco-bio-social conditions, community participation, political will and inter-sectorial partnership. Previously identified risk factors for persistent Triatoma dimidiata infestation include the presence of rodents and chickens, tiled roofs, dirt floors, partial wall plastering and
Corine Ngufor et al.
Malaria journal, 14, 464-464 (2015-11-20)
Novel indoor residual spraying (IRS) and long-lasting insecticidal net (LLIN) products aimed at improving the control of pyrethroid-resistant malaria vectors have to be evaluated in Phase II semi-field experimental studies against highly pyrethroid-resistant mosquitoes. To better understand their performance it

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service