Skip to Content
MilliporeSigma
All Photos(1)

Documents

PS1081

Chlorimuron ethyl

analytical standard

Synonym(s):

Ethyl 2-(4-chloro-6-methoxy-2-pyrimidinylcarbamoylsulfamoyl)benzoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H15ClN4O6S
CAS Number:
Molecular Weight:
414.82
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 100 mg

manufacturer/tradename

Chem Service, Inc. PS-1081

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCOC(=O)c1ccccc1S(=O)(=O)NC(=O)Nc2nc(Cl)cc(OC)n2

InChI

1S/C15H15ClN4O6S/c1-3-26-13(21)9-6-4-5-7-10(9)27(23,24)20-15(22)19-14-17-11(16)8-12(18-14)25-2/h4-8H,3H2,1-2H3,(H2,17,18,19,20,22)

InChI key

NSWAMPCUPHPTTC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David Carpenter et al.
Environmental pollution (Barking, Essex : 1987), 172, 275-282 (2012-11-10)
Current pesticide registration guidelines call for short-term testing of plants; long-term effects on vegetative parts and reproduction remain untested. The aims of our study were to determine level of recovery and recovery times for plants exposed to the sulfonylurea herbicide
Meie Wang et al.
Ecotoxicology and environmental safety, 72(8), 2121-2129 (2009-09-15)
It was observed in this work that joint effects of chlorimuron-ethyl and copper on chlorophyll content, the peroxidases (POD) activity, superoxide dismutases (SOD) activity and soluble protein content in leaves and roots of wheat were markedly significant. The chlorophyll content
Yu Sheng et al.
Ying yong sheng tai xue bao = The journal of applied ecology, 21(11), 2992-2996 (2011-03-03)
By using phospholipid fatty acids (PLFA) method, this paper studied the soil microbial community structure in the soybean fields with different chlorimuron-ethyl application history in Weihe district of Heilongjiang Province. In the meantime, the residual amount of chlorimuron-ethyl in soil
Xiaoli Zhang et al.
Environmental science and pollution research international, 18(3), 407-415 (2010-08-12)
Chlorimuron-ethyl has been widely used for the soybean production of China, but less information is available on the possible risk of long-term application of this herbicide. In this paper, soil samples were collected from the plots having been received 30 g
Jing Zhao et al.
Journal of agricultural and food chemistry, 54(14), 4948-4953 (2006-07-06)
Hybridomas secreting a monoclonal antibody (mAb) against the herbicide chlorimuron-ethyl (CE) were produced by fusing the mouse myeloma cell line (SP2/0) with splenocytes from a mouse immunized against the conjugate of the sulfonamide moiety of CE and bovine serum albumin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service