Skip to Content
MilliporeSigma
All Photos(1)

Documents

PS1068

Cypermethrin

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H19Cl2NO3
CAS Number:
Molecular Weight:
416.30
Beilstein:
8444118
EC Number:
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

packaging

ampule of 100 mg

manufacturer/tradename

Chem Service, Inc. PS-1068

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC1(C)C(\C=C(\Cl)Cl)C1C(=O)OC(C#N)c2cccc(Oc3ccccc3)c2

InChI

1S/C22H19Cl2NO3/c1-22(2)17(12-19(23)24)20(22)21(26)28-18(13-25)14-7-6-10-16(11-14)27-15-8-4-3-5-9-15/h3-12,17-18,20H,1-2H3

InChI key

KAATUXNTWXVJKI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 - STOT SE 3

Target Organs

Nervous system, Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

George P Lahm et al.
Bioorganic & medicinal chemistry letters, 15(22), 4898-4906 (2005-09-17)
A novel class of anthranilic diamides has been discovered with exceptional insecticidal activity on a range of Lepidoptera. These compounds have been found to exhibit their action by release of intracellular Ca2+ stores mediated by the ryanodine receptor. The discovery
Riin Muljar et al.
Journal of insect physiology, 58(11), 1469-1476 (2012-09-11)
Sublethal effects of pesticides in insects can be observed through physiological changes, which are commonly estimated by metabolic rate and respiratory patterns, more precisely by the patterns of discontinuous gas-exchange (DGE) cycles. The aim of the present research was to
Valentina Sechi et al.
Environmental pollution (Barking, Essex : 1987), 184, 586-596 (2013-11-10)
A soil multi-species, SMS, experimental test system consisting of the natural microbial community, five collembolan species and a predatory mite along with either Enchytraeus crypticus or the earthworm Eisenia fetida were exposed to α-cypermethrin. A comparison of the performance of
Hassane Adakal et al.
Experimental & applied acarology, 59(4), 483-491 (2012-09-13)
Traditional systems account for 95 % of the livestock produced in Burkina Faso. Tick infestation hampers livestock productivity in this area. However, little information exists on tick-control practices used by livestock farmers. We interviewed 60 stockbreeders working in traditional farming systems
M L Groner et al.
Journal of fish diseases, 36(3), 195-208 (2012-11-06)
Sea lice, Lepeophtheirus salmonis, are ectoparasites of farmed and wild salmonids. Infestations can result in significant morbidity and mortality of hosts in addition to being costly to control. Integrated pest management programmes have been developed to manage infestations, and in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service