Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHR9299

Supelco

4′-Methylacetophenone

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

Methyl p-tolyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4COCH3
CAS Number:
Molecular Weight:
134.18
Beilstein:
606053
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

Agency

(Not traceable to any primary pharmacopeia standard)

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

refractive index

n20/D 1.533 (lit.)

bp

226 °C (lit.)

mp

22-24 °C (lit.)

density

1.004 g/mL at 20 °C
1.005 g/mL at 25 °C (lit.)

application(s)

pharmaceutical

storage temp.

−20°C

SMILES string

CC(=O)c1ccc(C)cc1

InChI

1S/C9H10O/c1-7-3-5-9(6-4-7)8(2)10/h3-6H,1-2H3

InChI key

GNKZMNRKLCTJAY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ. Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate. Download your certificate at: http://www.sigma-aldrich.com

Application

These Reference Materials are suitable for use in several analytical applications including, but not limited to, method development for qualitative and quantitative analyses, daily calibration, and routine quality control testing.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service