Skip to Content
MilliporeSigma
All Photos(1)

Documents

PHL84267

Tubuloside A

phyproof® Reference Substance

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H48O21
CAS Number:
Molecular Weight:
828.76
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥90.0% (HPLC)

form

solid

manufacturer/tradename

PhytoLab

SMILES string

O=C(C)O[C@@H]1[C@@H](O[C@]2([H])O[C@@H](C)[C@H](O)[C@@H](O)[C@H]2O)[C@H](OC(/C=C/C3=CC=C(O)C(O)=C3)=O)[C@@H](CO[C@]4([H])O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O)O[C@H]1OCCC5=CC(O)=C(O)C=C5

Related Categories

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service