Skip to Content
MilliporeSigma
All Photos(1)

Documents

G005972

Mobile Phase Optimization in the Analysis of Lysozyme on Zenix® SEC-300

application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

HPLC: suitable

test parameters

column: Zenix SEC-300, 30 cm x 4.6 mm I.D., 3 μm particles (Z777028)
mobile phase: as indicated on figure
flow rate: 0.35 mL/min
temp.: 25 °C
detector: UV, 214 nm
injection: 5 μL
sample: lysozyme, 1 mg/mL

suitability

application for HPLC

application(s)

life science and biopharma

Legal Information

Zenix is a registered trademark of Sepax Technologies

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service