Skip to Content
MilliporeSigma
All Photos(1)

Documents

G005959

HPLC Analysis of Glycan and Isomers on Proteomix® SAX-NP5, High Resolution Separation

application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

HPLC: suitable

test parameters

column: Proteomix SAX-NP5, 15 cm x 4.6 mm I.D., 5 μm particles (Z777228)
mobile phase: (A) 2.5% (v/v) acetic acid, 0.5% TEA in water; 0.5% acetic acid in acetonitrile
gradient: 0-100% B in 60 min
flow rate: 0.3 mL/min
detector: Fluorescence, Ex/Em=360/425 nm
sample: 2-AA (anthranilic acid) labeled N-linked oligosaccharide profiling of an IgG1 sample

suitability

application for HPLC

application(s)

life science and biopharma

Legal Information

Proteomix is a registered trademark of Sepax Technologies

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service