Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G005953

HPLC Analysis of Fusion Protein on Zenix®-C SEC-300

application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

HPLC: suitable

test parameters

column: Zenix-C SEC-300, 30 cm x 7.8 mm I.D., 3 μm particles (Z777092)
mobile phase: 150 mM sodium phosphate buffer, pH 7.0 + 200 mM NaCl
flow rate: 1.0 mL/min
pressure: 1102 psi (76 bar)
column: 25 °C
detector: UV, 214 nm
injection: 10 μL
sample: 1 mg/mL fusion protein

suitability

application for HPLC

application(s)

life science and biopharma

Legal Information

Zenix is a registered trademark of Sepax Technologies

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service