Skip to Content
MilliporeSigma
All Photos(1)

Documents

G005912

LC/MS Analysis of Illicit Bath Salts on Ascentis® Express HILIC, Set 3

application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

HPLC: suitable

test parameters

column: Ascentis Express HILIC, 10 cm x 2.1 mm I.D., 2.7 μm particles (53939-U)
mobile phase: [A] 5 mM ammonium formate in acetonitrile; [B] water; (98:2, A:B)
flow rate: 0.6 mL/min
pressure: 267 psi (18.4 bar)
column temp.: 35 °C
detector: MS, ESI(+) TOF, Summed Ion Chromatogram
injection: 2 μL
sample: 500 ng/mL in acetonitrile

suitability

application for HPLC

application(s)

forensics and toxicology

Analysis Note

HILIC mode operation on an Ascentis Express column provided baseline resolution of these bath salts, including isobaric compounds. The highest quality LC-MS solvents were used to supply low background interference and low particulate contaminants for robust; trouble-free operation. Cerilliant CRMs provided reliable identification and quantification.

Legal Information

Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Pentedrone hydrochloride, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
    3-Fluoromethcathinone hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
    4-Methylethcathinone hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
    4-Fluoromethcathinone hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
    3,4-Dimethylmethcathinone hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service