Skip to Content
MilliporeSigma
All Photos(1)

Documents

G005878

LC/MS/MS Analysis of Vitamin D Metabolites on Ascentis® Express F5

application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

HPLC: suitable

test parameters

column: Ascentis Express  F5, 15 cm x 2.1 mm I.D., 2.7 μm particles (53571-U)
mobile phase: [A] 5 mM ammonium formate water; [B] methanol; (25:75, A:B)
flow rate: 0.4 mL/min
pressure: 5178 psi (357 bar)
column temp.: 40 °C
detector: ESI(+), 100-1000 m/z
injection: 2 μL, each compound 300 ng/mL in 40:60 (water:methanol)

suitability

application for HPLC

application(s)

clinical
vitamins, nutraceuticals, and natural products

Analysis Note

The metabolic role and health implications of the various vitamin D isoforms are of current clinical interest. Therefore, it is important to have an analytical method that will resolve all of the known isoforms with necessary sensitivity and specificity.This application shows an Ascentis Express F5 provided rapid resolution of the isobaric vitamin D metabolites. MS detection provides the necessary secondary resolution. The highest grade LC-MS solvents were used to supply low background interference and low particulate contaminants for robust, trouble-free operation. Cerilliant CRMs provided reliable identification and quantification.

Legal Information

Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    1α,25-Dihydroxyvitamin D3 solution, 5 μg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
    1α,25-Dihydroxyvitamin D2 solution, 5 μg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
    25-Hydroxyvitamin D3 solution, 100 μg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
    25-Hydroxyvitamin D2 solution, 50 μg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
    3-epi-25-Hydroxyvitamin D3, 98% (CP)
    3-epi-25-Hydroxyvitamin D2 solution, 100 μg/mL in ethanol, 98% (CP)
    1α-Hydroxyvitamin D3, ≥97.0% (HPLC)

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service