Skip to Content
MilliporeSigma
All Photos(1)

Documents

G005855

LC/MS Analysis of Antiarrhythmic Drugs and Metabolites on Ascentis® Express HILIC

application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

HPLC: suitable

test parameters

column: Ascentis Express HILIC, 10 cm x 2.1 mm I.D., 2.7 μm particles (53939-U)
mobile phase: [A] 5 mM ammonium formate; [B] 5 mM ammonium formate in acetonitrile, 5:95 (A:B), adjusted to pH 7.0 with formic acid
flow rate: 0.4 mL/min
pressure: 1305 psi (90 bar)
column temp.: 35 °C
detector: ESI+, 100-1000 m/z
injection: 0.5 μL
sample: each compound, 300 ng/mL in 75:25 (1% formic acid acetonitrile:water)

suitability

application for HPLC

application(s)

clinical

Analysis Note

The basic nature of these antiarrhythmic agents makes them ideal candidates for HILIC (hydrophilic interaction) chromatographic separation. The benefit of HILIC over traditional reversed-phase chromatography is two-fold for both sample introduction and analyte detection. First, the high acetonitrile concentration of HILIC mobile phases allows for direct analysis of precipitated plasma samples without the need for additional sample solvent exchange. Second, the high acetonitrile content provides increased analyte response in positive ESI MS detection. Of the various HILIC-mode columns tested, method development for this assay determined the Ascentis Express HILIC, 2.7 μm particles, provided the best chromatographic resolution of the antiarrhythmic drugs while maintaining high peak efficiency for enhanced detection levels.

Legal Information

Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Lidocaine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
    Amiodarone hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
    (±)-Flecainide solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
    N-Desethylamiodarone hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service